Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk8ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053777.1 | NP_446229.1 | 2 - 3 | 398 | 73 | |
GenBank mRNA | AF108959.1 | - | 1 - 2 | 336 | 73 | |
AF109772.1 | - | 1 - 2 | 245 | 73 | ||
AF109773.1 | - | 1 - 2 | 308 | 73 | ||
AF109774.1 | - | 2 - 3 | 398 | 73 | ||
DQ377223.1 | - | 1 - 2 | 208 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |