Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034948.1 | NP_001030120.1 | 3 - 4 | 314 | 71 | |
XM_006234786.2 | XP_006234848.1 | 3 - 4 | 250 | 71 | ||
XM_006234787.2 | XP_006234849.1 | 3 - 4 | 250 | 71 | ||
XM_008762106.1 | XP_008760328.1 | 3 - 4 | 314 | 71 | ||
GenBank mRNA | BC099795.1 | - | 3 - 4 | 314 | 7936116 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |