Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nutm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108957.1 | NP_001102427.1 | 4 - 5 | 1103 | 64 | |
XM_006234708.2 | XP_006234770.1 | 4 - 5 | 1099 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |