Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044247.1 | NP_001037712.1 | 6 - 7 | 884 | 81 | |
XM_006253100.2 | XP_006253162.1 | 6 - 7 | 2359 | 81 | ||
XM_008771246.1 | XP_008769468.1 | 6 - 7 | 1362 | 81 | ||
XM_008771247.1 | XP_008769469.1 | 6 - 7 | 957 | 81 | ||
XM_008771248.1 | XP_008769470.1 | 6 - 7 | 2202 | 81 | ||
GenBank mRNA | BC098935.1 | - | 6 - 7 | 884 | 7445963 | 81 |
FQ214804.1 | - | 5 - 6 | 744 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |