Search Thermo Fisher Scientific
Gene Symbol: Msr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191939.1 | NP_001178868.1 | 1 - 2 | 104 | 62 | |
XM_006253206.2 | XP_006253268.1 | 2 - 3 | 293 | 62 | ||
XM_006253207.2 | XP_006253269.1 | 2 - 3 | 288 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons |