Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013092.1 | NP_037224.1 | 3 - 4 | 384 | 68 | |
GenBank mRNA | D38495.1 | - | 3 - 4 | 384 | 68 | |
U67908.1 | - | 3 - 4 | 349 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |