Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dimt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106408.2 | NP_001099878.2 | 12 - 13 | 1120 | 117 | |
XM_006231902.2 | XP_006231964.1 | 12 - 13 | 1148 | 117 | ||
XM_006231903.2 | XP_006231965.1 | 13 - 14 | 1331 | 117 | ||
XM_006231905.2 | XP_006231967.1 | 11 - 12 | 1034 | 117 | ||
GenBank mRNA | BC158729.1 | - | 12 - 13 | 1120 | 9091721 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |