Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107889.1 | NP_001101359.1 | 6 - 7 | 800 | 119 | |
XM_006237363.2 | XP_006237425.1 | 7 - 8 | 790 | 119 | ||
XM_006237364.2 | XP_006237426.1 | 7 - 8 | 790 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |