Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044280.2 | NP_001037745.2 | 3 - 4 | 433 | 61 | |
GenBank mRNA | BC094517.1 | - | 3 - 4 | 378 | 7314749 | 61 |
CB726923.1 | - | 3 - 4 | 434 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |