Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053702.3 | NP_446154.3 | 4 - 5 | 853 | 107 | |
XM_006232266.2 | XP_006232328.1 | 5 - 6 | 990 | 107 | ||
XM_008760903.1 | XP_008759125.1 | 5 - 6 | 985 | 107 | ||
GenBank mRNA | AY623027.1 | - | 3 - 4 | 827 | 107 | |
AY623028.1 | - | 4 - 5 | 1232 | 107 | ||
BC127462.1 | - | 5 - 6 | 946 | 8373429 | 107 | |
CV114678.1 | - | 1 - 2 | 120 | 107 | ||
DY551463.1 | - | 4 - 5 | 564 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |