Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270039.1 | NP_001256968.1 | 3 - 4 | 1241 | 51 | |
XM_006231940.2 | XP_006232002.1 | 4 - 5 | 973 | 51 | ||
XM_008760737.1 | XP_008758959.1 | 5 - 6 | 1010 | 51 | ||
XM_008760738.1 | XP_008758960.1 | 4 - 5 | 979 | 51 | ||
GenBank mRNA | JQ013731.1 | - | 3 - 4 | 1241 | 51 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |