Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025647.1 | NP_001020818.1 | 11 - 12 | 1732 | 111 | |
XM_006231821.1 | XP_006231883.1 | 9 - 10 | 1428 | 111 | ||
XM_008760664.1 | XP_008758886.1 | 8 - 9 | 1473 | 111 | ||
XM_008760665.1 | XP_008758887.1 | 9 - 10 | 1351 | 111 | ||
GenBank mRNA | BC097489.1 | - | 11 - 12 | 1732 | 7460568 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |