Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsga13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107851.1 | NP_001101321.1 | 6 - 7 | 736 | 111 | |
XM_006236230.2 | XP_006236292.1 | 4 - 5 | 470 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |