Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053311.1 | NP_445763.1 | 11 - 12 | 2094 | 79 | |
XM_006241294.2 | XP_006241356.1 | 11 - 12 | 2186 | 79 | ||
XM_006241295.2 | XP_006241357.1 | 10 - 11 | 2063 | 79 | ||
XM_006241296.2 | XP_006241358.1 | 11 - 12 | 2186 | 79 | ||
GenBank mRNA | J03753.1 | - | 11 - 12 | 2094 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Transporter Rattus norvegicus Probe spans exons |