Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108094.2 | NP_001101564.1 | 7 - 8 | 969 | 97 | |
GenBank mRNA | BC128768.1 | - | 7 - 8 | 969 | 7125536 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |