Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Glt8d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765296.1 | XP_008763518.1 | 7 - 8 | 1267 | 81 | |
XM_008765297.1 | XP_008763519.1 | 7 - 8 | 1266 | 81 | ||
XM_008765298.1 | XP_008763520.1 | 7 - 8 | 1266 | 81 | ||
XM_008765299.1 | XP_008763521.1 | 7 - 8 | 1266 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |