Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198786.2 | NP_942081.2 | 2 - 3 | 457 | 77 | |
GenBank mRNA | AY079080.1 | - | 2 - 3 | 317 | 77 | |
AY079081.2 | - | 1 - 2 | 570 | 77 | ||
BC062388.1 | - | 2 - 3 | 457 | 5620891 | 77 |
Gene Symbol: LOC100911027 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748700.3 | XP_003748748.1 | 2 - 3 | 414 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |