Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133521.1 | NP_598205.1 | 2 - 3 | 209 | 102 | |
GenBank mRNA | AY059628.1 | - | 2 - 3 | 209 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |