Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173334.1 | NP_001166805.1 | 3 - 4 | 568 | 127 | |
XM_006241603.2 | XP_006241665.1 | 3 - 4 | 647 | 127 | ||
GenBank mRNA | BC166530.1 | - | 3 - 4 | 568 | 7189614 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |