Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam91a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127578.1 | NP_001121050.1 | 8 - 9 | 921 | 140 | |
XM_006241684.2 | XP_006241746.1 | 8 - 9 | 925 | 140 | ||
GenBank mRNA | BC166852.1 | - | 8 - 9 | 921 | 9092514 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |