Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012200.1 | NP_001012200.1 | 7 - 8 | 893 | 81 | |
XM_006242102.2 | XP_006242164.1 | 7 - 8 | 898 | 81 | ||
XM_006242103.2 | XP_006242165.1 | 7 - 8 | 944 | 81 | ||
XM_006242104.1 | XP_006242166.1 | 6 - 7 | 939 | 81 | ||
XM_008765764.1 | XP_008763986.1 | 6 - 7 | 850 | 81 | ||
GenBank mRNA | BC087669.1 | - | 7 - 8 | 893 | 7316813 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |