Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Far1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011933.1 | NP_001011933.1 | 7 - 8 | 953 | 65 | |
XM_006230017.2 | XP_006230079.1 | 8 - 9 | 1035 | 65 | ||
XM_006230018.2 | XP_006230080.1 | 8 - 9 | 960 | 65 | ||
XM_006230020.2 | XP_006230082.1 | 8 - 9 | 1037 | 65 | ||
GenBank mRNA | BC082015.1 | - | 7 - 8 | 953 | 7132096 | 65 |
FQ221385.1 | - | 8 - 9 | 1019 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |