Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glipr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011987.1 | NP_001011987.1 | 1 - 2 | 336 | 66 | |
GenBank mRNA | BC089858.1 | - | 1 - 2 | 336 | 7384037 | 66 |
FQ219892.1 | - | 1 - 2 | 341 | 66 | ||
FQ223414.1 | - | 1 - 2 | 341 | 66 | ||
FQ226490.1 | - | 1 - 2 | 357 | 66 | ||
FQ227021.1 | - | 1 - 2 | 356 | 66 | ||
FQ229493.1 | - | 1 - 2 | 363 | 66 | ||
FQ229810.1 | - | 1 - 2 | 402 | 66 | ||
FQ230706.1 | - | 1 - 2 | 364 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |