Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35e2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107998.1 | NP_001101468.1 | 5 - 6 | 1615 | 73 | |
XM_006239572.2 | XP_006239634.1 | 5 - 6 | 1177 | 73 | ||
XM_006239573.2 | XP_006239635.1 | 5 - 6 | 1294 | 73 | ||
XM_008764361.1 | XP_008762583.1 | 5 - 6 | 1294 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Rattus norvegicus Probe spans exons |