Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txlna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127633.1 | NP_001121105.1 | 4 - 5 | 717 | 70 | |
XM_006238972.2 | XP_006239034.1 | 4 - 5 | 724 | 70 | ||
GenBank mRNA | BC166486.1 | - | 4 - 5 | 717 | 7099983 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |