Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024306.1 | NP_001019477.1 | 3 - 4 | 644 | 137 | |
XM_006232006.2 | XP_006232068.1 | 3 - 4 | 1372 | 137 | ||
XM_008761519.1 | XP_008759741.1 | 3 - 4 | 1373 | 137 | ||
GenBank mRNA | BC079131.1 | - | 3 - 4 | 644 | 7127379 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |