Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Larp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044290.2 | NP_001037755.2 | 11 - 12 | 1649 | 92 | |
GenBank mRNA | BC083898.1 | - | 11 - 12 | 1586 | 7133376 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |