Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ntng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106465.1 | NP_001099935.1 | 3 - 4 | 1709 | 93 | |
XM_008761393.1 | XP_008759615.1 | 1 - 2 | 875 | 93 | ||
XM_008761394.1 | XP_008759616.1 | 1 - 2 | 875 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Inventoried Probe spans exons |