Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007653.1 | NP_001007654.1 | 7 - 8 | 709 | 65 | |
GenBank mRNA | BC083856.1 | - | 7 - 8 | 709 | 7125450 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |