Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748694.3 | XP_003748742.1 | 3 - 4 | 2362 | 88 | |
XM_006227909.2 | XP_006227971.1 | 3 - 4 | 2362 | 88 | ||
XM_006227910.2 | XP_006227972.1 | 3 - 4 | 2362 | 88 | ||
XM_006227911.2 | XP_006227973.1 | 3 - 4 | 2362 | 88 | ||
XM_008758741.1 | XP_008756963.1 | 3 - 4 | 2362 | 88 | ||
XM_008758742.1 | XP_008756964.1 | 3 - 4 | 2362 | 88 | ||
GenBank mRNA | BC169129.1 | - | 1 - 2 | 112 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |