Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024397.2 | NP_077373.2 | 6 - 7 | 780 | 64 | |
XM_006254353.2 | XP_006254415.1 | 6 - 7 | 851 | 64 | ||
XM_006254354.2 | XP_006254416.1 | 5 - 6 | 835 | 64 | ||
XM_006254355.2 | XP_006254417.1 | 5 - 6 | 835 | 64 | ||
XM_006254356.2 | XP_006254418.1 | 6 - 7 | 850 | 64 | ||
XM_006254357.2 | XP_006254419.1 | 5 - 6 | 835 | 64 | ||
XM_006254358.2 | XP_006254420.1 | 6 - 7 | 850 | 64 | ||
XM_006254359.2 | XP_006254421.1 | 5 - 6 | 835 | 64 | ||
XM_006254360.2 | XP_006254422.1 | 6 - 7 | 868 | 64 | ||
GenBank mRNA | AF176784.1 | - | 5 - 6 | 702 | 64 | |
BC129092.1 | - | 6 - 7 | 736 | 7192971 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |