Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npepo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008771429.1 | XP_008769651.1 | 5 - 6 | 1568 | 133 | |
XM_008771430.1 | XP_008769652.1 | 5 - 6 | 1568 | 133 | ||
XM_008771431.1 | XP_008769653.1 | 4 - 5 | 1415 | 133 | ||
XM_008771432.1 | XP_008769654.1 | 4 - 5 | 1407 | 133 | ||
XM_008771433.1 | XP_008769655.1 | 4 - 5 | 1280 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |