Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007558.1 | NP_001007559.1 | 875 | 89 | ||
XM_008766767.1 | XP_008764989.1 | 863 | 89 | |||
GenBank mRNA | AY686633.1 | - | 875 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Rattus norvegicus Probe spans exons |