Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004243.1 | NP_001004243.1 | 14 - 15 | 1806 | 156 | |
XM_008765690.1 | XP_008763912.1 | 15 - 16 | 2150 | 156 | ||
GenBank mRNA | BC079414.1 | - | 14 - 15 | 1806 | 7113336 | 156 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |