Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004243.1 | NP_001004243.1 | 4 - 5 | 565 | 91 | |
XM_008765690.1 | XP_008763912.1 | 5 - 6 | 909 | 91 | ||
GenBank mRNA | BC079414.1 | - | 4 - 5 | 565 | 7113336 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |