Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr89b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139486.1 | NP_001132958.1 | 4 - 5 | 410 | 114 | |
XM_006232976.2 | XP_006233038.1 | 2 - 3 | 399 | 114 | ||
GenBank mRNA | AY724489.1 | - | 4 - 5 | 341 | 114 | |
DY548897.1 | - | 4 - 5 | 410 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Rattus norvegicus Probe spans exons |