Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapre1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138509.3 | NP_612518.2 | 1 - 2 | 75 | 57 | |
GenBank mRNA | BC081726.1 | - | 1 - 2 | 75 | 7192469 | 57 |
U75920.1 | - | 1 - 2 | 43 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |