Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hm13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107789.1 | NP_001101259.1 | 6 - 7 | 779 | 69 | |
XM_006235268.1 | XP_006235330.1 | 6 - 7 | 790 | 69 | ||
XM_006235269.1 | XP_006235331.1 | 6 - 7 | 790 | 69 | ||
XM_008762330.1 | XP_008760552.1 | 6 - 7 | 790 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Rattus norvegicus Probe spans exons |