Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037657.1 | NP_001032746.1 | 145 | 109 | ||
GenBank mRNA | AY226099.1 | - | 145 | 109 |
Gene Symbol: LOC100910990 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006235453.2 | XP_006235515.1 | 1 - 2 | 173 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |