Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039004.1 | NP_001034093.1 | 1 - 2 | 189 | 123 | |
XM_006255132.2 | XP_006255194.1 | 1 - 2 | 822 | 123 | ||
GenBank mRNA | BC098748.1 | - | 1 - 2 | 189 | 7365565 | 123 |
BC168149.1 | - | 1 - 2 | 828 | 7457021 | 123 | |
FQ213930.1 | - | 1 - 2 | 232 | 123 | ||
FQ229498.1 | - | 1 - 2 | 230 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |