Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Dnase1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053907.1 | NP_446359.1 | 2 - 3 | 446 | 155 | |
GenBank mRNA | AF039852.1 | - | 2 - 3 | 396 | 155 | |
BC088122.1 | - | 2 - 3 | 444 | 7325326 | 155 | |
FQ226037.1 | - | 2 - 3 | 273 | 155 | ||
FQ227612.1 | - | 2 - 3 | 273 | 155 | ||
FQ230283.1 | - | 2 - 3 | 273 | 155 | ||
FQ230990.1 | - | 2 - 3 | 273 | 155 | ||
FQ231646.1 | - | 2 - 3 | 272 | 155 | ||
FQ231744.1 | - | 2 - 3 | 272 | 155 | ||
FQ231870.1 | - | 2 - 3 | 276 | 155 | ||
FQ231939.1 | - | 2 - 3 | 273 | 155 | ||
FQ233228.1 | - | 2 - 3 | 329 | 155 | ||
FQ234709.1 | - | 2 - 3 | 329 | 155 | ||
U75689.1 | - | 2 - 3 | 446 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Rattus norvegicus Probe spans exons |