Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fto | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039713.1 | NP_001034802.1 | 5 - 6 | 966 | 102 | |
XM_006255176.2 | XP_006255238.1 | 5 - 6 | 1399 | 102 | ||
XM_008772284.1 | XP_008770506.1 | 5 - 6 | 1295 | 102 | ||
XM_008772285.1 | XP_008770507.1 | 5 - 6 | 1297 | 102 | ||
GenBank mRNA | AM233906.1 | - | 5 - 6 | 966 | 102 | |
BC168239.1 | - | 5 - 6 | 981 | 5599736 | 102 | |
FQ211940.1 | - | 5 - 6 | 1006 | 102 | ||
FQ212297.1 | - | 5 - 6 | 995 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |