Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psma5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017282.1 | NP_058978.1 | 4 - 5 | 372 | 128 | |
GenBank mRNA | BC060575.1 | - | 4 - 5 | 393 | 6920479 | 128 |
D10756.1 | - | 4 - 5 | 372 | 128 | ||
FQ218317.1 | - | 4 - 5 | 389 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |