Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017093.1 | NP_058789.1 | 10 - 11 | 1241 | 62 | |
XM_008759108.1 | XP_008757330.1 | 11 - 12 | 3640 | 62 | ||
XM_008759109.1 | XP_008757331.1 | 10 - 11 | 1228 | 62 | ||
XM_008759110.1 | XP_008757332.1 | 10 - 11 | 3505 | 62 | ||
XM_008759111.1 | XP_008757333.1 | 10 - 11 | 1351 | 62 | ||
GenBank mRNA | D30041.1 | - | 10 - 11 | 1241 | 62 | |
DQ198085.1 | - | 9 - 10 | 960 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |