Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017093.1 | NP_058789.1 | 9 - 10 | 1117 | 84 | |
XM_008759108.1 | XP_008757330.1 | 10 - 11 | 3516 | 84 | ||
XM_008759109.1 | XP_008757331.1 | 9 - 10 | 1104 | 84 | ||
XM_008759110.1 | XP_008757332.1 | 9 - 10 | 3381 | 84 | ||
XM_008759111.1 | XP_008757333.1 | 9 - 10 | 1227 | 84 | ||
GenBank mRNA | D30041.1 | - | 9 - 10 | 1117 | 84 | |
DQ198085.1 | - | 8 - 9 | 836 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |