Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106628.1 | NP_001100098.1 | 1 - 2 | 140 | 51 | |
XM_006237677.1 | XP_006237739.1 | 1 - 2 | 151 | 51 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |