Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgke | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039341.1 | NP_001034430.1 | 9 - 10 | 1616 | 88 | |
XM_006247070.2 | XP_006247132.1 | 9 - 10 | 1811 | 88 | ||
XM_006247071.2 | XP_006247133.1 | 8 - 9 | 1739 | 88 | ||
GenBank mRNA | BC110049.1 | - | 9 - 10 | 1616 | 7316890 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |