Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080149.1 | NP_001073618.1 | 7 - 8 | 784 | 147 | |
XM_006252765.2 | XP_006252827.1 | 8 - 9 | 857 | 147 | ||
XM_006252767.2 | XP_006252829.1 | 8 - 9 | 824 | 147 | ||
XM_008771099.1 | XP_008769321.1 | 8 - 9 | 810 | 147 | ||
XM_008771100.1 | XP_008769322.1 | 8 - 9 | 857 | 147 | ||
GenBank mRNA | BC129117.1 | - | 7 - 8 | 784 | 7114504 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Phosphatase 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |