Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grid2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105910.1 | NP_001099380.1 | 2 - 3 | 319 | 115 | |
XM_006248865.2 | XP_006248927.1 | 2 - 3 | 389 | 115 | ||
XM_006248866.2 | XP_006248928.1 | 3 - 4 | 981 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |