Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007732.1 | NP_001007733.1 | 3 - 4 | 469 | 109 | |
XM_006253872.2 | XP_006253934.1 | 3 - 4 | 438 | 109 | ||
GenBank mRNA | BC079063.1 | - | 3 - 4 | 469 | 7121006 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |